missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex NHP RANTES (CCL5) Simplex

Catalog No. 501125745
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125745 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125745 Supplier Invitrogen™ Supplier No. EPX01A40287901

Includes

  • concentrated Magnetic Beads coated with monoclonal antibody to non-human primate Rantes
  • non-human primate Rantes Standard (lyophilized)
  • Biotin-Conjugate (50x) anti-non-human primate Rantes monoclonal antibody
  • Universal Assay Buffer 10x

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • It is recommended to dilute RANTES samples when measured in serum or plasma
  • The recommended sample dilution factor is 1:50

Specifications

Quantity 96 Tests
Species Monkey
For Use With (Application) Multiplex Immunoassay
Protein Subtype CCL5 (RANTES)
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL
Synonym Small inducible cytokine A5, SIS-delta