missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex NHP Chemokine Panel (10 plex)
Description
Includes
- NHP Standard Mix A
- NHP Bead Mix C
- NHP Detection Antibody Mix C
- Reading Buffer
- Wash Buffer 10x
- Streptavidin-PE
- Universal Assay Buffer
- Detection Antibody Diluent
- PCR 8-Tube Strip
- Adhesive Film
- Greiner Microplate black 96 well
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
- This kit does not contain a Multiscreen Filter Plate
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Monkey |
| For Use With (Application) | Multiplex Immunoassay |
| Protein Subtype | CCL11 (Eotaxin), CCL2 (MCP-1), CCL4 (MIP-1 beta), CXCL10 (IP-10), CXCL12 alpha (SDF-1 alpha), CXCL13, CXCL9 (MIG), I-TAC/CXCL11, IL-8 (CXCL8), MIP-1 alpha |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Sample Volume | Serum, Plasma: 25 μL; CCS: 50 μL |