missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex Mouse IL-15/IL-15R Simplex

Catalog No. 501125665
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125665 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125665 Supplier Invitrogen™ Supplier No. EPX01A26023901

Includes

  • concentrated Magnetic Beads coated with monoclonal antibody to mouse IL-15/IL-15R
  • mouse IL-15/IL-15R Standard (lyophilized)
  • mouse Biotin-Conjugate anti-mouse IL-15/IL-15R monoclonal antibody

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins

Specifications

Quantity 96 Tests
Species Mouse
For Use With (Application) Multiplex Immunoassay
Protein Subtype CD215 (IL-15 Receptor alpha), IL-15, IL-15/IL-15R complex
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL
Synonym Interleukin 15, Interleukin 15 Receptor