missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Mouse ENA-78 (CXCL5) Simplex
Description
Includes
- concentrated Magnetic Beads coated with monoclonal antibody to mouse ENA-78
- mouse ENA-78 Standard (lyophilized)
- mouse Biotin-Conjugate (50x) anti-mouse ENA-78 monoclonal antibody
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Mouse |
| For Use With (Application) | Multiplex Immunoassay |
| Sample Type | Cell Culture Supernatants, Serum, Plasma |
| Synonym | SCYB5 |