missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Mouse 36plex Control Set
Description
eBioscience™ ProcartaPlex Mouse Cytokine & Chemokine Panel 1A (35 plex) (Mfr. Part No. EPX360-26092-901) is the ideal solution for assessing multiple protein biomarkers in a single sample. The assays can quantitate multiple targets in cell/tissue culture supernatants, serum and plasma using Luminex™ xMAP technology for the detection of secreted proteins including cytokines, chemokines, growth factors and other proteins.
Specifications
Specifications
| Product Type | Multiplex Control Set |
| Quantity | 1 Pk. |
| Species | Mouse |
| For Use With (Application) | Multiplex Immunoassay |
| For Use With (Equipment) | Luminex™ Instruments |
| Product Line | ProcartaPlex |
| Protein Subtype | CCL2 (MCP-1), CCL5 (RANTES), CCL7 (MCP-3), CXCL1 (GRO alpha), CXCL10 (IP-10), CXCL2 (MIP-2), ENA-78, Eotaxin, G-CSF, GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-10, IL-12, IL-13, IL-15/IL-15R complex, IL-17A, IL-18, IL-2, IL-22, IL-23, IL-27, IL-28, IL-3, IL-31, IL-4, IL-5, IL-6, IL-9, LIF, M-CSF, MIP-1 alpha, MIP-1 beta, TNF alpha |