missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Human Viral Reagent Kit DFS Item

Designed to be used with the Coronavirus Simplex kits to combine single analytes into a multiplex assay

Supplier:  Invitrogen™ EPX01010430901

Catalog No. EPX01010430


Only null left

Description

Description

  • The ProcartaPlex Viral Reagent Kit is designed to be used with the Coronavirus Simplex kits to combine single analytes into a multiplex assay.
  • This provides maximum flexibility to design assay panels as needed. This kit includes all reagents except for the target-specific beads (sold separately) to run the assay.
  • ProcartaPlex immunoassays are an ideal solution for assessing multiple protein biomarkers in a single sample.
  • Suitable sample types: serum, plasma
  • Sample volume: serum, plasma: 25 μL
  • Reported application: multiplex immunoassay
Specifications

Specifications

Basic and Setup Kits
Human
See Certificate of Analysis
ProcartaPlex
Wet Ice
96 Tests
Combinable
Luminex Instruments

• 1 vial Biotinylated Detection Antibody Mix (50X)
• 2 vials Human Control-High
• 2 vials Human Control-Medium
• 2 vials Human Control-Low
• 1 bottle Reading Buffer (1X)
• 1 bottle Wash Buffer (10X)
• 1 bottle Assay Buffer (1X)
• 1 bottle Detection Antibody Diluent (1X)
• 8-tube strip
• Adhesive film
• Flat bottom 96-well plate, black

Store at 2°C to 8°C.

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.