missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human TRAIL-R2 Simplex
Description
Includes
- concentrated Magnetic Beads coated with monoclonal antibody to human TRAIL-R2
- human TRAIL-R2 Standard (lyophilized)
- Biotin-Conjugate (50x) anti-human TRAIL-R2 monoclonal antibody
ProcartaPlex™ assays are subject to the same stringent and robust development and manufacturing as the extensively validated Platinum and Instant ELISA kits. Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma.
- Reactivity/Species: Human
- Suitable Sample Types: Cell culture supernatant, serum, plasma
- Sample Volume: Serum, Plasma—25μL; CS—50μL
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Human |
| For Use With (Application) | Multiplex Immunoassay |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Sample Volume | Serum, Plasma: 25 μL; CCS: 50 μL |
| Synonym | Death receptor 5, TNFRSF10B, CD262, DR5 |