missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex Human TIM-3 (CD366) Simplex

Catalog No. 501125633
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125633 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125633 Supplier Invitrogen™ Supplier No. EPX01A12219901

Includes

  • concentrated Magnetic Beads coated with monoclonal antibody to human TIM-3 (CD366)
  • human TIM-3 (CD366) Standard (lyophilized)
  • Biotin-Conjugate (50x) anti-human TIM-3 (CD366) monoclonal antibody

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins

Specifications

Quantity 96 Tests
Species Human
For Use With (Application) Multiplex Immunoassay
Protein Subtype CD366 (TIM3)
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL
Synonym HAVCR2, T cell immunoglobulin and mucin domain-containing protein 3