missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex Human Th1/Th2 & Chemokine Panel 1 (20 plex)

Catalog No. 501125847
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125847 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125847 Supplier Invitrogen™ Supplier No. EPX20012173901

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

2 vials Human Standard Mix A

2 vials Human Standard Mix B

1 vial Human Bead Mix B

1 vial Human Bead Mix E

1 vial Human Detection Antibody Mix B

1 vial Human Detection Antibody Mix E

1 bottle Reading Buffer

1 bottle Wash Buffer 10x

1 bottle Streptavidin-PE

1 bottle Universal Assay Buffer

1 bottle Detection Antibody Diluent

PCR 8-Tube Strip

Adhesive Film

Greiner Microplate 96 well

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • This kit does not contain a Multiscreen Filter Plate
  • Suitable Samples: cell culture supernatant, serum, plasma

Specifications

Applications Multiplex
Host Species Human
Quantity 96 Tests