missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human RANKL Simplex
Description
Includes
- concentrated Magnetic Beads coated with monoclonal antibody to human RANKL
- human RANKL Standard (lyophilized)
- Biotin-Conjugate (50x) anti-human RANKL monoclonal antibody
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Human |
| For Use With (Application) | Multiplex Immunoassay |
| Protein Subtype | CD254 (RANK Ligand) |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Sample Volume | Serum, Plasma: 25 μL; CCS: 50 μL |
| Synonym | Rankl-related activation-induced cytokine,CD254, ODF, OPGL, TRANCE, TNFSF11 |