missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEK1 Human ProcartaPlex™ Simplex Kit

Catalog No. 502884744
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502884744 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502884744 Supplier Invitrogen™ Supplier No. EPX01097014901
Only null left

Invitrogen ProcartaPlex Human MEK1 Simplex Kit enables precise analysis of the Protein of Interest.

The ProcartaPlex Human MEK1 Simplex Kit measures the level of total MEK1 in cell lysates. It is designed to be combinable with other Simplex kits so you can create your own multiplex panel that utilizes Luminex xMAP technology for protein quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

For preparation of cell lysates, we recommend the use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-99999-000) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease and Phosphatase Inhibitor Single to Use Cocktail 100X (Cat. No. 78440 or 78442). These buffers are not included with the kit. Depending on your cell system, other buffers can be suitable as well.

The ProcartaPlex Human MEK1 Simplex Kit is part of the ProcartaPlex Human MAPK Pathway Panel (total) 6plex (Cat. No. EPX060-97017-901), which complements the ProcartaPlex Human MAPK Pathway Panel (phospho) 6plex (Cat. No. EPX060-97117-PHO).

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • More results per sample - measure multiple protein targets simultaneously in a single 25 to 50 μL sample
  • Well-established Luminex technology - highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25 to 50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin to R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

Specifications

Product Type Simplex Kit
Quantity 96 Tests
Species Human
Combinability Combinable
Bead Type Magnetic
Bead Region MEK1 [63]
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
For Use With (Equipment) Luminex™ Instruments
Gene MAP2K1
Gene Alias CFC3, MAPKK1, MEK1, MKK1, PRKMK1, mitogen-activated protein kinase kinase 1, MEL
Gene ID (Entrez) 5604
Gene Symbol MAP2K1
High-throughput Compatibility Yes
Interassay CV See Certificate of Analysis
Intraassay CV See Certificate of Analysis
Product Line ProcartaPlex
Sample Type Cell lysates
Sample Volume Cell lysates (prediluted): 25 μL
UniProt ID Q02750
Synonym Mitogen-activated protein kinase
Content And Storage 1 vial Capture Beads (50x)
1 vial biotinylated Detection Antibody (50x)
2 vials Standard Mix (lyophilized)

Store as indicated on the Certificate of Analysis.
Shipping Condition Wet Ice
Research Area Signaling
Show More Show Less

For Research Use Only.