missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex Human LAP (TGF beta 1) Simplex

Catalog No. 501125482
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125482 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125482 Supplier Invitrogen™ Supplier No. EPX01012065901

Includes

  • concentrated Magnetic Beads coated with monoclonal antibody to human LAP
  • human LAP Standard (lyophilized)
  • Biotin-Conjugate (50x) anti-human LAP monoclonal antibody

Assess multiple protein biomarkers in a single sample. Our ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins.

  • ProcartaPlex™ assays are subject to the same stringent and robust development and manufacturing as are our extensively validated Platinum and Instant ELISA kits
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma

Specifications

Quantity 96 Tests
Species Human
For Use With (Application) Multiplex Immunoassay
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL
Synonym Latency Associated Peptide