Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human HER-2 Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Supplier: Invitrogen™ EPX01A10207901
Includes:
- concentrated Magnetic Beads coated with monoclonal antibody to human HER-2
- human HER-2 Standard (lyophilized)
- Biotin-Conjugate (50x) anti-human HER-2 monoclonal antibody
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
| 96 Tests | |
| Multiplex Immunoassay | |
| Serum, Plasma: 25 μL; CCS: 50 μL |
| Human | |
| Cell Culture Supernatant, Plasma, Serum | |
| ERBB2, Herstatin, CD340 |