missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex Human Essential Panel 2 (4 plex)

Catalog No. 501125801
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125801 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125801 Supplier Invitrogen™ Supplier No. EPX04010008901

Includes

  • Human Standard Mix A
  • Human Standard Mix B
  • Human Essentail Panel 2 Bead Mix
  • Human Essentail Panel 2 Detection Antibody Mix
  • Reading Buffer
  • Wash Buffer 10x
  • Streptavidin-PE
  • Universal Assay Buffer
  • Detection Antibody Diluent
  • PCR 8-Tube Strip
  • Adhesive Film
  • Greiner Microplate black 96 well

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • This kit does not contain a Multiscreen Filter Plate

Specifications

Quantity 96 Tests
Species Human
For Use With (Application) Multiplex Immunoassay
Protein Subtype IL-1 beta, IL-10, IL-17A, IL-8 (CXCL8)
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL