missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human Coagulation Panel 3 (4 plex)
Description
Includes
- Human Coagulation Panel 3 Reference Plasma Standard
- Human Coagulaton Panel 3 Bead Mix
- Human Coagulation Panel 3 Detection Antibody Mix
- Reading Buffer
- Wash Buffer 10x
- Streptavidin-PE
- Universal Assay Buffer 10X
- Detection Antibody Diluent
- PCR 8-Tube Strip
- Adhesive Film
- Greiner Microplate black 96 well
- The human Coagulation panels are designed for the use with plasma samples (EDTA, Citrate)
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
- This kit does not contain a Multiscreen Filter Plate
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Human |
| For Use With (Application) | Multiplex Immunoassay |
| Protein Subtype | Factor IX, Protein C, Protein S, Von Willebrand Factor |
| Sample Type | Plasma (EDTA, Citrate) |
| Sample Volume | Plasma (Citrat, EDTA): 25 μL |