missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex Human 45plex Control Set

Catalog No. 501125863
Change view
Click to view available options
Quantity:
1 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125863 1 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125863 Supplier Invitrogen™ Supplier No. EPX45012171CTR

Contains a high and low control for all analytes. eBioscience™ ProcartaPlex Human 45plex Control Set is used with the ProcartaPlex Human Cytokine/Chemokine/Growth Factor Panel 1 (45 plex).

eBioscience™ ProcartaPlex Human Cytokine/Chemokine/Growth Factor Panel 1 (45 plex) (Mfr. Part No. EPX450-12171-901) is the ideal solution for assessing multiple protein biomarkers in a single sample. The assays can quantitate multiple targets in cell/tissue culture supernatants, serum and plasma using Luminex™ xMAP technology for the detection of secreted proteins including cytokines, chemokines, growth factors and other proteins.

Specifications

Product Type Multiplex Control Set
Quantity 1 Pk.
Species Human
For Use With (Application) Multiplex Immunoassay
For Use With (Equipment) Luminex™ Instruments
Product Line ProcartaPlex
Protein Subtype Brain-Derived Neurotrophic Factor (BDNF), CCL2 (MCP-1), CCL3 (MIP-1 alpha), CCL5 (RANTES), CXCL1 (GRO alpha), CXCL10 (IP-10), CXCL12 alpha (SDF-1 alpha), EGF, Eotaxin, FGF-2, GM-CSF, HGF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1 Receptor Antagonist, IL-10, IL-12, IL-13, IL-15, IL-17A, IL-18, IL-2, IL-21, IL-22, IL-23, IL-27, IL-31, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-9, LIF, MIP-1 beta, NGF beta, PDGF-BB, PIGF-1, Stem Cell Factor (SCF), TNF alpha, TNF beta, VEGF-A, VEGF-D