missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human 4-1BBL (CD137L) Simplex
Description
Includes
- concentrated Magnetic Beads coated with monoclonal antibody to human 4-1BBL (CD137L)
- human 4-1BBL (CD137L) Standard (lyophilized)
- Biotin-Conjugate (50x) anti-human 4-1BBL (CD137L) monoclonal antibody
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Human |
| For Use With (Application) | Multiplex Immunoassay |
| Protein Subtype | CD137 Ligand (4-1BB Ligand) |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Sample Volume | Serum, Plasma: 25 μL; CCS: 50 μL |
| Synonym | TNFSF9, 4-1BB Ligand, CD137 Ligand |