Learn More
Thermo Scientific™ Procarbazine hydrochloride
Supplier: Thermo Scientific™ 461180010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Procarbazine hydrochloride | |
| 2% max. | |
| C12H19N3O·HCl | |
| procarbazine hydrochloride, procarbazine hcl, matulane, nathulane, natunalar, mih hydrochloride, pcb hydrochloride, procarbazine.hcl, ibenzmethyzine hydrochloride, natulan hydrochloride | |
| CC(C)NC(=O)C1=CC=C(C=C1)CNNC.Cl | |
| 257.76 | |
| CHEBI:71428 | |
| ≥97.5% (HPLC) |
| 366-70-1 | |
| Authentic | |
| 1g | |
| DERJYEZSLHIUKF-UHFFFAOYSA-N | |
| 4-[(2-methylhydrazinyl)methyl]-N-propan-2-ylbenzamide;hydrochloride | |
| 9703 | |
| 257.76 |
Chemical Identifiers
| 366-70-1 | |
| 257.76 | |
| procarbazine hydrochloride, procarbazine hcl, matulane, nathulane, natunalar, mih hydrochloride, pcb hydrochloride, procarbazine.hcl, ibenzmethyzine hydrochloride, natulan hydrochloride | |
| CHEBI:71428 | |
| CC(C)NC(=O)C1=CC=C(C=C1)CNNC.Cl |
| C12H19N3O·HCl | |
| DERJYEZSLHIUKF-UHFFFAOYSA-N | |
| 9703 | |
| 4-[(2-methylhydrazinyl)methyl]-N-propan-2-ylbenzamide;hydrochloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause cancer.
Suspected of causing genetic defects.
May damage fertility or the unborn child.
GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
EINECSNumber : 206-678-6
RUO â Research Use Only