missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Procarbazine hydrochloride

Catalog No. p-7077874
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461182500 250 mg
AC461180010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461182500 Supplier Thermo Scientific Chemicals Supplier No. 461182500
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 366-70-1
Molecular Formula C12H19N3O·HCl
Molecular Weight (g/mol) 257.76
InChI Key DERJYEZSLHIUKF-UHFFFAOYSA-N
Synonym procarbazine hydrochloride, procarbazine hcl, matulane, nathulane, natunalar, mih hydrochloride, pcb hydrochloride, procarbazine.hcl, ibenzmethyzine hydrochloride, natulan hydrochloride
PubChem CID 9703
ChEBI CHEBI:71428
IUPAC Name 4-[(2-methylhydrazinyl)methyl]-N-propan-2-ylbenzamide;hydrochloride
SMILES CC(C)NC(=O)C1=CC=C(C=C1)CNNC.Cl

Specifications

Loss on Drying 2% max.
Infrared Spectrum Authentic
Quantity 250 mg
Formula Weight 257.76
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Procarbazine hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause cancer.
Suspected of causing genetic defects.
May damage fertility or the unborn child.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 206-678-6

RUO – Research Use Only