missing translation for 'onlineSavingsMsg'
Learn More

Procaine hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019567
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019567 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019567 Supplier Merck Emd Millipore Supplier No. PHR11611G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Procaine hydrochloride is a synthetic compound that belongs to the class of local anesthetic drugs. It has a structure similar to the natural compounds that participate in nerve impulse transmission. It shows interaction with the cellular membranes causing membrane lipids to expand, membrane-bound calcium to displace and change the structure of the proteins that are responsible for regulating the cell permeability and shape.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 51-05-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula H2 NC6H4 CO2 CH2 CH2 N(C2H5)2·HCl
MDL Number MFCD00013000
Melting Point 155°C to 156°C
Synonym 4-Aminobenzoic acid 2-diethylaminoethyl ester; p-Aminobenzoic acid diethylaminoethyl ester hydrochloride; Novocaine hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 272.77
Grade Certified Reference Material
Show More Show Less