missing translation for 'onlineSavingsMsg'
Learn More

Probucol, MilliporeSigma™ Supelco™

Catalog No. 111013615
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013615 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013615 Supplier Merck Emd Millipore Supplier No. P967250G
Only null left

Probucol is a diphenolic compound possessing antioxidant and anti-inflammatory properties. It is commercially used as a lipid lowering drug, which helps in reducing the total plasma cholesterol, low density lipoprotein cholesterol (LDL) and high density lipoprotein cholesterol (HDL).

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 23288-49-5
Physical Form Neat
Linear Formula C31H48O2S2
MDL Number MFCD00079281
Type Analytical Standard
Quantity 50 g
Formula Weight 516.84
Grade Analytical Standard