missing translation for 'onlineSavingsMsg'
Learn More

Probenecid 98.0+%, TCI America™

Supplier:  TCI America P1975100G

 View more versions of this product

Catalog No. P1975100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Probenecid
200°C
C13H19NO4S
100 g
DBABZHXKTCFAPX-UHFFFAOYSA-N
4-(dipropylsulfamoyl)benzoic acid
4911
285.36
Crystalline Powder
57-66-9
White
MFCD00038402
probenecid, 4-dipropylsulfamoyl benzoic acid, probenecid acid, probecid, benecid, benemid, benuryl, probalan, probenemid, probexin
CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(O)=O
285.36
CHEBI:8426
≥98.0% (HPLC,T)

Chemical Identifiers

57-66-9
285.36
DBABZHXKTCFAPX-UHFFFAOYSA-N
4911
4-(dipropylsulfamoyl)benzoic acid
C13H19NO4S
MFCD00038402
probenecid, 4-dipropylsulfamoyl benzoic acid, probenecid acid, probecid, benecid, benemid, benuryl, probalan, probenemid, probexin
CHEBI:8426
CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DG9400000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions