missing translation for 'onlineSavingsMsg'
Learn More

Proadifen hydrochloride

Catalog No. p-7042808
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63833ME 500 mg
AAJ6383303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63833ME Supplier Thermo Scientific Chemicals Supplier No. J63833ME

CAS: 62-68-0 | C23H32ClNO2 | 389.964 g/mol

Proadifen hydrochloride is an inhibitor of NOS1, AChR, cytochrome P-450, and KIR6. Proadifen hydrochloride bind to the protein moiety of P-450, also reduced the hypoxic response. of Proadifen hydrochloride have good antiarrhythmic effects. SKF-525A substantially reduced the depletion of cardiac norepinephrine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Proadifen hydrochloride is an inhibitor of NOS1, AChR, cytochrome P-450, and KIR6. Proadifen hydrochloride bind to the protein moiety of P-450, also reduced the hypoxic response. of Proadifen hydrochloride have good antiarrhythmic effects. SKF-525A substantially reduced the depletion of cardiac norepinephrine.

Solubility
Soluble in methanol or water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

Chemical Identifiers

CAS 62-68-0
Molecular Formula C23H32ClNO2
Molecular Weight (g/mol) 389.964
MDL Number MFCD00055151
InChI Key FHIKZROVIDCMJA-UHFFFAOYSA-N
Synonym proadifen hydrochloride, proadifen hcl, propyladiphenin, skf-525a hydrochloride, skf 525a, sk&f 525-a, proadifen hydrochloride usan, skf 525-a, skf-525a, hydrochloride
PubChem CID 65341
IUPAC Name 2-(diethylamino)ethyl 2,2-diphenylpentanoate;hydrochloride
SMILES CCCC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)OCCN(CC)CC.Cl

Specifications

Melting Point 122°C to 123°C
Color White
Quantity 500 mg
Solubility Information Soluble in methanol or water
Formula Weight 389.97
Physical Form Powder
Chemical Name or Material Proadifen hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber YV7175000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only