missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proadifen hydrochloride, ≥95%, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS | 62-68-0 |
| Physical Form | Neat |
| Linear Formula | C23H31 NO2 · HCl |
| MDL Number | MFCD00055151 |
| Synonym | alpha-Phenyl-alpha-propylbenzeneacetic acid 2-(diethylamino)ethyl ester; N,N-Diethylaminoethyl 2,2-diphenylvalerate; SKF-525 A |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 389.96 |
| Percent Purity | ≥95% |
| Show More |