missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pro-Detect™ Rapid DYKDDDDK-His Assay Kit

Catalog No. PIA38500
Change view
Click to view available options
Quantity:
10 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA38500 10 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA38500 Supplier Thermo Scientific™ Supplier No. A38500
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

10 assays

Pro-Detect™ Rapid Assay Kit is a single-use, dipstick lateral flow kit for quick, easy detection of tagged proteins in protein expression and purification systems

  • Lateral flow assay uses membrane-based strips coated with dye-bound conjugated antibodies specific to the tag to provide a visual, color readout for detection within ten minutes.
  • Detect proteins directly from cell culture media (supernatant) or cell lysates without any special instrumentation or handling.
  • Lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
  • Dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5 to 10 minutes
  • Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentrations
  • Specific—assay does not cross-react with common lysis and protein preparation reagents
  • Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
  • Long shelf life—stable for at least one year at 4°C

Specifications

Content And Storage • 10 Rapid Assay Strips
• Sample Diluent, 15 mL

Sufficient for 10 assay determinations

Store at 2–8°C.
Detectable Analytes DYKDDDDK-His (FLAG-His) double tagged proteins
Assay Lateral Flow Rapid Assay
Detection Method Chromogenic
Product Line Pro-Detect
Product Type Assay
Specificity Protein tag
Sufficient For 10 Assays
For Use With (Application) Lateral Flow Rapid Assay
Quantity 10 Assays
Shipping Condition Wet Ice
Target Protein
Show More Show Less
Can I use the Pro-Detect assay kits for rapidly isotyping antibodies?

The Pro-Detect Rapid Antibody Isotyping Kit is a lateral-flow assay with high sensitivity for rapid determination of the immunoglobulin class, subclass, and light chain type of monoclonal antibodies. Two different formats of rapid isotyping kits are available. These include dipstick format and cassette-based format. One can use these kits to identify the isotype of a mouse, rat, or human antibody. Make sure to follow the recommended dilution guidelines.

I am using a Pro-Detect assay kit and cannot differentiate banding pattern between control lysates and lysates that are actually expressing proteins of interest. What is the problem?

1) It is likely that there could be a substance interfering with the assay. Check the compatibility table on this page (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/pro-detect-rapid-assays.html), under “Detergent and salt compatibility limits”.
2) The expression level of the actual tagged protein is below the detectable range of the assay.

Using Pro-Detect assay kits, can I detect the protein of interest in a mixture of proteins such as a crude lysate?

Yes, first, make sure that the protein of interest is tagged and that we offer a Pro-Detect assay kit that can detect that tag. Secondly, make sure that you follow the guidelines to detect proteins in crude lysate and if necessary increase or decrease the lysate concentration to bring the protein of interest within the detectable range of the assay.

Note that the guidelines for the recommended detection ranges reflect the concentration of the pure tagged protein in a given sample mixture.

How do I interpret the gradual disappearance of four lines in the Pro-Detect competition assay kits?

As the concentration of the tag in the protein sample increases, the number of lines will decrease until they all disappear. The concentration of the tagged proteins is inversely related to the number of lines appearing on the strip.

Are the results of Pro-Detect assay kits comparable to western blotting?

Yes. Western blotting is a widely used and accepted tool to identify a specific protein in a complex mixture. However, it is a time-consuming and labor-intensive process, with many preparative steps and time intervals to manage. The Pro-Detect rapid assays are able to provide comparable accuracy but with much shorter processing time (10-15 min) and less hands-on steps. For further information, please see the data shown on this page (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/pro-detect-rapid-assays.html) under “Results are comparable to western blotting”.

How specific are the Pro-Detect assay kits? Is there any cross-reactivity observed among various kits ?

Except for MBP Pro-Detect strips which cross-react with bacterial proteins, no other cross-reactivity of Pro-Detect strips has been observed. An example is shown in the figure on this page (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/pro-detect-rapid-assays.html) under “No cross-reactivity with other targets” where there was no cross-reactivity of Pro-Detect rapid GST assay strips with proteins endogenous to mammalian, bacterial, Sf9 cells (lanes 1-5) or with any other non-specific tag proteins (lanes 7-19). In addition, even when using 100% of Minimum Essential Medium with 10% FBS, there was no cross-reactivity detected.

Are the Pro-Detect assay kits compatible with common detergent and salt concentrations used in protein purification/detection protocols?

We examined the performance of the Pro-Detect rapid assay in the presence of various concentrations of detergents and salts that are commonly used in protein expression and isolation. Reagents tested include NaCl, Urea, Triton-X 100, SDS, NP40, EDTA, Glycerol, KCl, CHAPS, RIPA, BPER, and MPER. For estimates of each assay tolerance level for the tested detergents and salts, please refer to the table under “Detergent and salt compatibility limits” at https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/pro-detect-rapid-assays.html

What is the ideal range of protein concentration needed for using Pro-Detect assay kits?

For sandwich assays, the range is 0.1-10 µg/mL of pure tagged protein.
Note: The lateral flow assays can detect outside of this range; however, intensity of the bands will be lower at concentrations higher and lower than the working range. It is not recommended to use at concentrations less than 0.01 µg/mL. In a mixture of proteins such as lysates, the target protein has to be in the detectable range (0.1-10 µg/mL) for the assay to be successful.

For competition assays, the range is 4-20 µg/ml.
Note: The competitive lateral flow assays can detect down to 1 µg/mL, but loss of band intensity may be less distinguished. As there is no upper limit, the lateral flow strip will continue to show positive results at concentrations greater than 20 µg/mL, visualized by complete loss of test lines (test lines closest to the sample will disappear first). It is not recommended to use at concentrations less than 1 µg/mL.


For Research Use Only. Not for use in diagnostic procedures.