missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRMT7 Recombinant Rabbit Monoclonal Antibody (23GB2380)

Catalog No. MA553549
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553549 20 μL
MA553548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553549 Supplier Invitrogen™ Supplier No. MA553549
Only null left

Rabbit Recombinant Monoclonal Antibody

PRMT7 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Arginine methylation is an irreversible post translational modification which has been implicated in RNA processing and trafficking, receptor-mediated signaling, and transcription. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Outside of these domains, epitopes were identified and antibodies against all 7 PRMT members have been developed.

Specifications

Antigen PRMT7
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2380
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRMT7
Gene Accession No. Q5U4E8, Q922X9, Q9NVM4
Gene Alias [Myelin basic protein]-arginine N-methyltransferase PRMT7; 4933402B05Rik; BC006705; FLJ10640; Histone-arginine N-methyltransferase PRMT7; KIAA1933; myelin basic protein-arginine N-methyltransferase; PRMT7; PRMT-7; protein arginine methyltransferase 7; protein arginine N-methyltransferase 7; RGD1304869
Gene Symbols PRMT7
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRMT7 (1-38AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214572, 361402, 54496
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less