missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRMT3 Recombinant Rabbit Monoclonal Antibody (23H2L13)
Description
This antibody is predicted to react with Mouse, Bovine.
Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins.
Specifications
Specifications
| Antigen | PRMT3 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23H2L13 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2-7.4 |
| Gene | Prmt3 |
| Gene Accession No. | O60678, O70467 |
| Gene Alias | 2010005E20Rik; 2410018A17Rik; AL033309; heterogeneous nuclear ribonucleoprotein methyltransferase-like 3; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3; HMT1 hnRNP methyltransferase-like 3; Hrmt1l3; PRMT3; protein arginine methyltransferase 3; protein arginine N-methyltransferase 3; protein arginine N-methyltransferase 3(hnRNP methyltransferase S. cerevisiae)-like 3 |
| Show More |