missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149851
Description
PRM2 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| PRM2 | |
| Polyclonal | |
| Unconjugated | |
| PRM2 | |
| AI528784; Basic nuclear protein HPI1; Basic nuclear protein HPI2; Basic nuclear protein HPS1; Basic nuclear protein HPS2; cancer/testis antigen family 94, member 2; CT94.2; P2'; P2''; PP2-A; PP2-B; PP2-C; PP2-D; Prm2; Prm-2; protamine 2; Protamine mP2; Protamine-2; Sperm histone HP2; Sperm histone HP3; Sperm histone HP4; Sperm histone P2; sperm protamine P2; Sperm protamine P3; Sperm protamine P4; sperm protamine-P2; testicular secretory protein Li 40 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5620 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P04554 | |
| PRM2 | |
| Recombinant Protamine 2 (PRM2) (Val2-Arg100). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |