Learn More
Thermo Scientific™ Printers for CryoMed™ and CryoPlus™ Freezers
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with this optional printer.
Supplier: Thermo Scientific™ 4000665
Description
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with Thermo Scientific™ Thermal Printers for CryoMed and CryoPlus™ Freezers. These printers are available in 120V or 220V models, and include cable assembly and RS-232 connector kit.
- Model 4000565 - 120V, 60Hz printer with cable assembly and RS-232 connector kit
- Model 4000665 - 220V, 50Hz printer with cable assembly and RS-232 connector kit
- Model 4000566 - Thermal printer paper (5 rolls/package)
Chemical Identifiers
| All CryoMed Controlled-Rate Freezers |
Specifications
| Thermal Data Printer | |
| All CryoMed Controlled-Rate Freezers | |
| Warranty varies by country, please contact us for details | |
| 220 V |