Learn More
Gibco™ Primary Mouse Cortical Neurons
Accelerate your neuroscience research with convenient, easy-to-use Gibco Primary Mouse Cortical Neurons. High viability post-thaw, high purity, high quality.
$1,040.00 - $1,288.00
Specifications
| Content And Storage | Store in liquid nitrogen, vapor-phase. |
|---|---|
| Age | Fetal |
| Cell Type | Neural Cells |
| Culture Type | Mammalian Cell Culture |
| Form | Cryopreserved |
Description
Gibco Primary Mouse Cortical Neurons are isolated and cryopreserved from the brain cortices of C57BL/6 embryonic day-17 mice. The cryopreserved cortical neurons offer convenience and flexibility compared to primary neuron isolation workflows, and the availability of large single lots provides greater consistency to cell-based assays. The Primary Mouse Cortical Neurons are ready-to-use and provide reliable quality. All product lots yield post-thaw viability in the range of 50-80%, have high neuronal purity, and are verified for functionality.
- High viability—the cryopreserved cortical neurons exhibit exceptional survival rates (50% or greater), enabling reliable and consistent results
- High purity—immunofluorescence staining conducted on every lot with MAP2 to confirm high neuronal purity
- Dependable quality—release tests include sterility testing and mycoplasma testing
- Functionally validated—product demonstrates long-term survival and neurotransmitter response
Culture Gibco Primary Mouse Cortical Neurons with the B-27 Plus Neuronal Culture System to achieve the most robust performance.
Specifications
| Store in liquid nitrogen, vapor-phase. | |
| Neural Cells | |
| Cryopreserved | |
| Cortex | |
| Mouse Cortical Neuron | |
| Sterility, Viability, Mycoplasma |
| Fetal | |
| Mammalian Cell Culture | |
| Mouse | |
| 1 Million | |
| Mouse strain is C57BL/6 |
For Research Use Only. Not for use in diagnostic procedures.