missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Primary Mouse Cortical Neurons

Catalog No. A15586
Encompass
Change view
Click to view available options
Quantity:
1 x 106
4 x 106
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15586 4 x 106
A15585 1 x 106
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15586 Supplier Gibco™ Supplier No. A15586

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Accelerate your neuroscience research with convenient, easy-to-use Gibco Primary Mouse Cortical Neurons. High viability post-thaw, high purity, high quality.

Gibco Primary Mouse Cortical Neurons are isolated and cryopreserved from the brain cortices of C57BL/6 embryonic day-17 mice. The cryopreserved cortical neurons offer convenience and flexibility compared to primary neuron isolation workflows, and the availability of large single lots provides greater consistency to cell-based assays. The Primary Mouse Cortical Neurons are ready-to-use and provide reliable quality. All product lots yield post-thaw viability in the range of 50-80%, have high neuronal purity, and are verified for functionality.

  • High viability—the cryopreserved cortical neurons exhibit exceptional survival rates (50% or greater), enabling reliable and consistent results
  • High purity—immunofluorescence staining conducted on every lot with MAP2 to confirm high neuronal purity
  • Dependable quality—release tests include sterility testing and mycoplasma testing
  • Functionally validated—product demonstrates long-term survival and neurotransmitter response

Culture Gibco Primary Mouse Cortical Neurons with the B-27 Plus Neuronal Culture System to achieve the most robust performance.

Specifications

Content And Storage Store in liquid nitrogen, vapor-phase.
Age Fetal
Cell Type Neural Cells
Culture Type Mammalian Cell Culture
Form Cryopreserved
Species Mouse
Tissue Cortex
No. of Cells 1 Million
Product Type Mouse Cortical Neuron
Quantity 4 x 106
Remarks Mouse strain is C57BL/6
Tested For Sterility, Viability, Mycoplasma
Show More Show Less
Would I be able to find out the sex of our Primary Mouse Cortical Neurons?

Primary Mouse Cortical Neurons are mixed sex, because it's too hard to identify the sex of a mouse fetus.

I'm currently culturing neurons. Which media system do you recommend using in conjunction with the Poly-D-Lysine matrix?

We recommend using the B-27 Plus Neuronal Culture System when culturing neurons on Poly-D-Lysine. The B-27 Plus Neuronal Culture System is a complete media system that includes the B-27 Plus Supplement and the Neurobasal Plus Medium.

What cell types will Poly-D-Lysine support as an attachment matrix?

The Poly-D-Lysine matrix has successfully supported the attachment of primary rodent neurons, iPSC-derived neurons and PC12 cells.

For Research Use Only. Not for use in diagnostic procedures.