missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Primary Hepatocyte Thawing and Plating Supplements

Catalog No. CM3000
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CM3000 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CM3000 Supplier Gibco™ Supplier No. CM3000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Primary Hepatocyte Thawing and Plating Supplements

Hepatocyte Thawing and Plating Supplement Pack

Hepatocyte Thawing and Plating Supplement Packs allow you to supplement up to 500 ml of Williams Medium E without phenol red (or a suitable alternative basal medium) for the purpose of plating fresh or cryopreserved hepatocytes.

Hepatocyte Thawing and Plating Supplement Packs contain:

  • Prequalified fetal bovine serum
  • Dexamethasone
  • A cocktail solution of fetal bovine serum, penicillin-streptomycin, human recombinant insulin, GlutaMAX™, and HEPES

    For research use only. Not intended for human or animal therapeutic or diagnostic use.

    Related Links:

    Learn more about our cell culture reagents for hepatocytes.

    Learn more about our cryopreserved hepatocytes.

    Learn about our full line of hepatocytes.
  • Specifications

    Content And Storage Store Cocktail A and dexamethasone in refrigerator (2–8°C) and protect from light.
    Store FBS in freezer (-5 to -20°C) and protect from light.

    Cell Type Hepatocyte Cells
    Form Cryopreserved
    Product Type Hepatocyte Supplement
    With Additives GlutaMAX, HEPES
    Without Additives No Phenol Red
    Product Line Gibco
    Quantity 1 kit

    For Research Use Only. Not for use in diagnostic procedures.