missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Primary Hepatocyte Maintenance Supplements

Catalog No. CM4000
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CM4000 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CM4000 Supplier Gibco™ Supplier No. CM4000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Primary Hepatocyte Maintenance Supplements

Hepatocyte Maintenance Supplement Pack

Hepatocyte Maintenance Supplement Packs allow you to supplement up to 500 ml of Williams Medium E without phenol red (or suitable alternative basal medium) for the purpose of incubating hepatocytes in suspension or plated cultures.

Hepatocyte Maintenance Supplement Packs contain:

  • Dexamethasone
  • A cocktail solution of penicillin-streptomycin, ITS+ (insulin, transferrin, selenium complex, BSA, and linoleic acid), GlutaMAX™, and HEPES

    For research use only. Not intended for human or animal therapeutic or diagnostic use.

    Related Links:

    Learn more about our cell culture reagents for hepatocytes.

    Learn more about our cryopreserved hepatocytes.

    Learn about our full line of hepatocytes.
  • Specifications

    Content And Storage Store Cocktail B and dexamethasone in refrigerator (2–8°C) and protect from light.
    Cell Type Hepatocyte Cells
    Culture Type Mammalian Cell Culture, Insect Cell Culture
    Form Cryopreserved
    Product Type Hepatocyte Supplement
    With Additives GlutaMAX, HEPES
    Without Additives No Phenol Red
    Product Line Gibco
    Quantity 1 kit

    For Research Use Only. Not for use in diagnostic procedures.