Learn More
Primary Drinking Water Metals without Mercury for Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier:  SPEX CertiPrep Group LLC EP8500N
 View more versions of this product
Catalog No. EP8500N
Only null left
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
AnalyteConcentration
Ba100μg/mL
Ag10μg/mL
As10μg/mL
Cr10μg/mL
Pb10μg/mL
Cd5μg/mL
Se5μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
Primary Drinking Water Metals without Mercury
Triple-leached LDPE bottle
AA, ICP
Certified Reference Material
500 mL
Various
Drinking Water Pollutant Standard
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
2% HNO3
Environmental
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Product intended for laboratory use only