missing translation for 'onlineSavingsMsg'
Learn More

Primary Drinking Water Metals For Environmental Analysis, SPEX CertiPrep™

Catalog No. EP8
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EP8 125 mL
EP8500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EP8 Supplier SPEX CertiPrep Group LLC Supplier No. EP8

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ba100μg/mL
Ag10μg/mL
As10μg/mL
Cr10μg/mL
Pb10μg/mL
Cd5μg/mL
Se5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Primary Drinking Water Metals
Packaging Triple-leached LDPE bottle
Name Note Drinking Water Pollutant Standard
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Quantity 125 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Various
Show More Show Less

Product intended for laboratory use only