Learn More
Genlantis™ PrimaPure™ Human Dermal Microvascular Endothelial Cells (HMVEC) and Kits
Functionally characterized
Supplier: Genlantis™ PH10005A
Description
HMVEC are isolated from normal human neonatal foreskin or adult skin capillaries. They are cryopreserved at second passage and can be cultured and propagated at least 16 population doublings. They are responsive to cytokine stimulation in the expression of cell adhesion molecules. HMVEC express CD36 constitutively, while large vessel endothelial cells do not. HMVEC is an organ-specific endothelial cell system for the study of angiogenesis1, tumor metastasis2,3, wound healing4, inflammation5,6, macromolecule transport7, and drug metabolism8. There is growing evidence of differences in physiology and pharmacology between microvascular endothelial cells and macrovascular endothelial cells9,10.
- Single donors to allow variability comparisons
- Each lot tested negative for HIV, Hepatitis B and C, mycoplasma, bacteria, yeast, and fungi
- Performance guaranteed with Genlantis media and reagents
1. Swerlick, R.A. et.al, J. Invest. Dermatol. 97(2):190 (1991).
2. Folkman, J. et al, Nature 288:551 (1980).
3. Lee, K.H. et al, J. Invest. Dermatol. 98(1):79 (1992).
4. Deryugina, E.I. et al, Hybridoma 15(4):279 (1996).
5. Sepp, N.T. et al, J. Invest. Dermatol. 104(2):277 (1995).
6. Swerlick, R.A. et al, J. Invest. Dermatol. 100(1):111S (1993).
7. Hanasaki, K. et al, J. Bio. Chem. 270(13):7533 (1995).
8. Pruckler, J.M. et al, Pathobiology 61:283 (1993).
9. Foster, C.A. et al, J. Dermatol. 21(11):847 (1994).
10. Schnitzer, J.E. et al, BBRC. 199(1):11 (1994).
Specifications
| Adult HDMEC | |
| HMVEC Growth Medium | |
| Human Neonatal Foreskin or Adult Skin |
| For the study of angiogenesis, tumor metastasis, wound healing, inflammation, macromolecule transport, drug metabolism | |
| >500000 Cells | |
| 16 Population Doublings |