missing translation for 'onlineSavingsMsg'
Learn More

Prima-1, Thermo Scientific Chemicals

A cell-permeable quinuclidinone analog

Supplier:  Thermo Scientific Chemicals J64071MA

 View more versions of this product

Catalog No. AAJ64071MA


Only null left
Explore available promotions

Description

Description

A cell-permeable quinuclidinone analog.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Prima-1
White
MFCD04974196
Light sensitive
Soluble in water at 10mg/ml
C1CN2CCC1C(=O)C2(CO)CO
185.223
185.2
5608-24-2
C9H15NO3
10 mg
prima-1, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2, octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-3-quinuclidinone, p53 reactivation and induction of massive apoptosis, 2,2-bishydroxymethyl-1-aza-bicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2 octan-3-one, prima 1, tocris-1862, prima 1;prima1
RFBVBRVVOPAAFS-UHFFFAOYSA-N
2,2-bis(hydroxymethyl)-1-azabicyclo[2.2.2]octan-3-one
322968
Solid

Chemical Identifiers

5608-24-2
185.223
RFBVBRVVOPAAFS-UHFFFAOYSA-N
322968
C1CN2CCC1C(=O)C2(CO)CO
C9H15NO3
MFCD04974196
prima-1, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2, octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-3-quinuclidinone, p53 reactivation and induction of massive apoptosis, 2,2-bishydroxymethyl-1-aza-bicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2 octan-3-one, prima 1, tocris-1862, prima 1;prima1
2,2-bis(hydroxymethyl)-1-azabicyclo[2.2.2]octan-3-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only