missing translation for 'onlineSavingsMsg'
Learn More

Prilocaine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7135370
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
P20381G 1 g
P20385G 5 g
2 options

Catalog No. P20381G

Supplier: TCI America P20381G

Only null left

Chemical Identifiers

CAS 1786-81-8
Molecular Formula C13H21ClN2O
Molecular Weight (g/mol) 256.774
MDL Number MFCD00079279
InChI Key BJPJNTKRKALCPP-UHFFFAOYSA-N
Synonym prilocaine hydrochloride, propitocaine hydrochloride, prilocaine hcl, xylonest, prilocaine chloride, citanest hydrochloride, citanest plain, n-2-methylphenyl-2-propylamino propanamide hydrochloride, 2-propylamino
PubChem CID 92163
ChEBI CHEBI:32053
IUPAC Name N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride
SMILES CCCNC(C)C(=O)NC1=CC=CC=C1C.Cl

Specifications

Melting Point 166°C
Color White
Quantity 1 g
Formula Weight 256.77
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Prilocaine Hydrochloride
RTECSNumber UG5775000
TSCA No