missing translation for 'onlineSavingsMsg'
Learn More

Presenilin-1 CT Mouse anti-Mouse, Rat, Clone: PS1-loop, MilliporeSigma™

Catalog No. MAB5232I25
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5232I25 25 μL
MAB5232I100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB5232I25 Supplier MilliporeSigma Supplier No. MAB5232I25UL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Presenilin-1 CT Monoclonal antibody specifically detects Presenilin-1 CT in Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting.

Specifications

Antigen Presenilin-1 CT
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PS1-loop
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen A recombinant fragment corresponding to 116 amino acids from the loop fusion region (263-378) in the C-terminal half of human Presenilin-1.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less