missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Striated Skeletal Muscle (Frog)

Catalog No. S30619
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30619 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30619 Supplier Eisco™ Supplier No. BS18270

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prepared microscope slide of frog striated skeletal muscle. Stained for better visualization of characteristic morphology and structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic morphology and structures

Specifications

Description Frog Striated Skeletal muscle stained for better visualization of characteristic morphology and structures
Classification Zoology
Color Clear
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1 Ea.
Specimen Type Frog Striated Skeletal Muscle
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less