missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Frog Ovary, H&E Stain

Catalog No. S30655
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30655 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30655 Supplier Eisco™ Supplier No. BS18299

Prepared microscope slide of frog ovary stained with hematoxylin and eosin. Ovary is a part of female reproductive system. Showing characteristic structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures

Specifications

Description Frog Ovary showing characteristic structures
Classification Reproductive System
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Species Female Mammal
Specimen Type Frog Ovary
Stain Type H and E stain
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less