missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Prepared Microscope Slide - Spinal Cord Cross Section

Catalog No. S29645
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S29645 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29645 Supplier Eisco™ Supplier No. BS18030

Prepared microscope slide of spinal cord

  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization
  • Ideal for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows characteristic anatomy of spinal cord
  • Cross section

Specifications

Biosafety Level 1
Concentration 10%
Description Microscope Slide
Classification Cell Biology
Color Clear
Material Glass
Mounting Type Cross Section
Type Microbiology Slide
For Use With (Application) Microscopy
Grade 9 to 12
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Quantity 1
Species Rabbit
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less