missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Prepared Microscope Slide - Motor Nerve Ending Plates, Whole Mount

Catalog No. S29644
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S29644 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29644 Supplier Eisco™ Supplier No. BS18029

Prepared microscope slide of motor nerve ending plates

  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization
  • Ideal for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows characteristic structures of motor nerve ending plates
  • Wholemount
  • Motor nerve end plates function to establish synaptic contact with muscle fibers

Specifications

Biosafety Level 1
Concentration 10%
Description Microscope Slide
Classification Microbiology
Color Clear
Material Glass
Mounting Type Cross Section
Type Microbiology Slide
For Use With (Application) Microscopy
Grade 9 to 12
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Quantity 1
Species Rabbit
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less