missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Prepared Microscope Slide - Medium Artery and Vein, Cross-Section

Catalog No. S29633
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S29633 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. S29633 missing translation for 'mfr' Eisco™ missing translation for 'supplierNo' BS18018

Prepared microscope slide of a cross section of medium artery and vein

  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization
  • Ideal for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows characteristic structures of artery and vein anatomy
  • Ideal for classrooms studying anatomy

Specifications

Biosafety Level 1
Concentration 10%
Description Microscope Slide
Classification Cell Biology
Color Clear
Material Glass
Mounting Type Cross Section
Type Microbiology Slide
For Use With (Application) Microscopy
Grade 9 to 12
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Quantity 1
Species Human
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less