missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Lilium Stem Combination, Logitudinal Section &Cross Section

Catalog No. S30648
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30648 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30648 Supplier Eisco™ Supplier No. BS18292

Prepared composite microscope slide of a cross section and longtitudinal section of lilium stem. Lilium is a monocot. Stained for better visualization of characteristic monocot structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures of monocot stem in two different points of view

Specifications

Description Cross and longitudinal sections of Lilium stem stained for better visualization of characteristic monocot structures
Classification Botany
Color Clear
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1 Ea.
Specimen Type Lilium Stem Combination
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less