missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Tilia 2 Yr Stem (Basswood), Cross Section

Catalog No. S30670
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30670 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30670 Supplier Eisco™ Supplier No. BS18318

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prepared microscope slide of a cross section of two year old tilia stem. Tilia, commonly known as basswood, is a dicot. Stained for better visualization of characteristic structures including secondary xylem. Great choice for biology classrooms to explore structure function relationships.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures including secondary xylem

Specifications

Description Cross section of Tilia (Basswood) stem, stained for better visualization of characteristic structures including secondary xylem
Classification Botany
Color Clear
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1 Ea.
Specimen Type Tilia Stem (2 Years Old)
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less