missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Tilia Old Stem (Basswood), Cross Section

Catalog No. S30605
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30605 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30605 Supplier Eisco™ Supplier No. BS18264

Prepared microscope slide of Tilia old stem. Tilia is commonly known as basswood. Stained for better visualization of characteristic structures, including secondary xylem annual rings. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures, including secondary xylem annual rings

Specifications

Description Cross Section of Tilia (Basswood) Old Stem shows characteristic structures such as secondary xylem annual rings
Classification General
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Tilia Old Stem (Basswood)
Size 1 x 3 in.
Slide Type Prepared Slide