missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Spirogyra Scalariform, Whole Mount
Description
- Pre-cleaned glass slide with ground edges
- Size 1 x 3 in. (25 x 77 mm)
- Professionally stained samples for best visualization
- Great choice for biology classrooms to explore structure-function relationships per NGSSs standards
- Showing conjugation canals as well as the characteristic spiral chloroplast arrangement
Specifications
Specifications
| Description | Scalariform conjugation is a form of sexual reproduction where filaments are joined via conjugation tubes. Whole Mount shows conjugation canals as well as the characteristic spiral chloroplast arrangement. |
| Classification | Reproductive System |
| Material | Glass |
| Mounting Type | Whole Mount |
| Type | Prepared Microscope Slide |
| For Use With (Application) | Microscopy |
| Grade | 9 to 12 |
| Quantity | 1 Ea. |
| Specimen Type | Spirogyra Scalariform Conjugation |
| Size | 1 x 3 in. |
| Show More |