missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Gymnosperm Angiosperm Stem

Catalog No. S30646
Change view
Click to view available options
Gymnosperm Angiosperm Stem
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S30646 Gymnosperm Angiosperm Stem
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30646 Supplier Eisco™ Supplier No. BS18290

Prepared microscope slide of gymnosperm and angiosperm stem section. Angiosperms are plants which have flowers and fruits, while gymnosperms have no flowers and fruits and have unenclosed seeds. Great choice for biology classrooms to examine the stems of gymnosperms and angiosperms side by side.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures

Specifications

Description Gymnosperm and Angiosperm stem sections. Angiosperms are plants which have flowers and fruits. Gymnosperms have no flowers or fruits, and have unenclosed seeds.
Classification Botany
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Gymnosperm Angiosperm Stem
Size 1 x 3 in.
Slide Type Prepared Slide