missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Herbaceous Roots, Cross Section

Catalog No. S30623
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30623 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30623 Supplier Eisco™ Supplier No. BS18274

Prepared microscope slide of a cross section of herbaceous roots. Stained for better visualization of characteristic structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures

Specifications

Description Cross Section of herbaceous roots, stained for better visualization of characteristic structures
Classification General
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Herbaceous Roots
Size 1 x 3 in.
Slide Type Prepared Slide